| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900010400.mm.1 |
NM_145496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010400.mm.1 |
NM_145496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010400.mm.1 |
TC1900001109.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900010401.mm.1 |
NM_145496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010401.mm.1 |
NM_145496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010401.mm.1 |
TC1900001109.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900010399.mm.1 |
NM_145496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010399.mm.1 |
NM_145496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010399.mm.1 |
TC1900001109.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900010402.mm.1 |
NM_145496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010402.mm.1 |
NM_145496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900010402.mm.1 |
TC1900001109.mm.1 |
MTA TC |