Assay Details
| Gene Symbol: Tkfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145496.1 | NP_663471.1 | 5 - 6 | 653 | 68 | |
| GenBank mRNA | AK138956.1 | - | 693 | 68 | ||
| BC021917.1 | - | 5 - 6 | 653 | 4481611 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tkfc
Genomic Map