Assay Details
| Gene Symbol: Wasf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145155.3 | NP_660137.1 | 1 - 2 | 333 | 68 | |
| GenBank mRNA | AK044481.1 | - | 354 | 68 | ||
| BC027038.1 | - | 1 - 2 | 333 | 5363785 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044481.1, BC027038.1, NM_145155.3 transcripts.
Set Membership: |
|

Wasf3
Genomic Map