Assay Details
| Gene Symbol: Sh3d19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082414.2 | NP_001075883.2 | 1 - 2 | 921 | 90 | |
| GenBank mRNA | BC028254.1 | - | 1 - 2 | 469 | 90 | |
| D89677.2 | - | 7 - 8 | 1407 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D89677.2 transcripts.
Set Membership: |
|

Sh3d19
Genomic Map