Assay Details
Gene Symbol: 4930556G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040655.1 | 6 - 7 | 745 | 123 | ||
GenBank mRNA | AK016143.1 | - | 6 - 7 | 746 | 123 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Eph receptor A7 | Epha7 |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
-1.9mb | Sense | Chr. 4 - 28813044 - 28967511 on NCBI Build GRCm38 |
Downstream | mitogen-activated protein kinase kinase kinase 7 | Map3k7 |
B430101B05, C87327, Tak1 |
1.1mb | Sense | Chr. 4 - 31962296 - 32023470 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670262 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38222 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 553291 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8365 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796630 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806831 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973345 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498026 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47619 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726647 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191143 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715418 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169332 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454663 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720581 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566533 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700275 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697148 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14119 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476525 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78843 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880086 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416242 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199455 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643463 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374408 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175592 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301479 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810097 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666095 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658017 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84315 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57026 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741111 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936186 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334925 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92826 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416506 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714976 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960006 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858907 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759004 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149239 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362498 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3919 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47100 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153646 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96953 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276425 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212969 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483792 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357326 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181474 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136056 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862078 | BC029684.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862078 | XM_205950.7 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862078 | XM_913671.3 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955348 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823093 | AK031627.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823093 | BC139770.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823093 | NM_009044.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823093 | X15842.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823093 | X60271.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160847 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178002 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370720 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501032 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134939 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136161 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14732 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174314 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113063 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63974 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139186 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16208 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35514 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166981 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42841 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00841335 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190871 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05191051 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05191005 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190909 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190778 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190946 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05191075 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00841294 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00841267 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05191093 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190819 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190835 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190979 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190894 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05190940 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05191028 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109125 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05190979 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05191051 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05190894 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05190778 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05190835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03382585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03469724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03469722 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812487 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811739 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03469696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812197 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812143 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03580579 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03752981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811855 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812360 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03557482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812212 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03558090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03721160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812105 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03403432 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03823071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03573339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03287918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03752985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811994 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811867 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03469716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812111 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03469715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812198 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03876919 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812113 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03876917 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03224576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03954692 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03469694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03823070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03580577 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03377932 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22812041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03520000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22811985 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0400001506.mm.1 | NR_040655.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400001506.mm.1 | NR_040655.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400001506.mm.1 | TC0400000244.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 359478 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105670072 | - | - |
