Assay Details
| Gene Symbol: Mms22l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199467.2 | NP_955761.2 | 8 - 9 | 1383 | 146 | |
| GenBank mRNA | AK089531.1 | - | 1 - 2 | 123 | 146 | |
| AK144799.1 | - | 8 - 9 | 1302 | 146 | ||
| AK166072.1 | - | 8 - 9 | 1394 | 146 | ||
| AY512923.1 | - | 3 - 4 | 517 | 146 | ||
| BC094429.1 | - | 7 - 8 | 727 | 146 | ||
| BC137910.1 | - | 3 - 4 | 293 | 8860929 | 146 | |
| BC145028.1 | - | 3 - 4 | 293 | 9053576 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mms22l
Genomic Map