Assay Details
| Gene Symbol: Mms22l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199467.2 | NP_955761.2 | 11 - 12 | 1744 | 132 | |
| GenBank mRNA | AK166072.1 | - | 11 - 12 | 1755 | 132 | |
| AY512923.1 | - | 6 - 7 | 878 | 132 | ||
| BC051674.1 | - | 2 - 3 | 191 | 132 | ||
| BC094429.1 | - | 10 - 11 | 1088 | 132 | ||
| BC137910.1 | - | 6 - 7 | 654 | 8860929 | 132 | |
| BC145028.1 | - | 6 - 7 | 654 | 9053576 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mms22l
Genomic Map