Assay Details
| Gene Symbol: Fhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021318.3 | NP_067293.1 | 1 - 2 | 169 | 87 | |
| GenBank mRNA | AF083394.1 | - | 1 - 2 | 117 | 87 | |
| AK006414.1 | - | 1 - 2 | 158 | 87 | ||
| BC048473.1 | - | 1 - 2 | 169 | 6770515 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF083394.1, AK006414.1, BC048473.1, NM_021318.3 transcripts.
Set Membership: |
|

Fhl5
Genomic Map