Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 10 - 11 | 1222 | 145 | |
| GenBank mRNA | AK007993.2 | - | 10 - 11 | 1167 | 145 | |
| AK013382.1 | - | 3 - 4 | 485 | 145 | ||
| AK032663.1 | - | 10 - 11 | 1158 | 145 | ||
| AK076206.1 | - | 10 - 11 | 1163 | 145 | ||
| AK129211.1 | - | 9 - 10 | 1135 | 145 | ||
| AK132697.1 | - | 10 - 11 | 1186 | 145 | ||
| BC058809.1 | - | 9 - 10 | 2546 | 145 | ||
| BC080203.1 | - | 10 - 11 | 1200 | 145 | ||
| BC099529.1 | - | 10 - 11 | 1209 | 145 | ||
| BC132195.1 | - | 10 - 11 | 1174 | 40130472 | 145 | |
| BC138153.1 | - | 10 - 11 | 1174 | 8861172 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013382.1 transcripts.
Set Membership: |
|

Ufl1
Genomic Map