Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 11 - 12 | 1345 | 137 | |
| GenBank mRNA | AK007993.2 | - | 11 - 12 | 1290 | 137 | |
| AK013382.1 | - | 4 - 5 | 608 | 137 | ||
| AK032663.1 | - | 11 - 12 | 1281 | 137 | ||
| AK076206.1 | - | 11 - 12 | 1286 | 137 | ||
| AK129211.1 | - | 10 - 11 | 1258 | 137 | ||
| AK132697.1 | - | 11 - 12 | 1309 | 137 | ||
| BC058809.1 | - | 10 - 11 | 2669 | 137 | ||
| BC080203.1 | - | 11 - 12 | 1323 | 137 | ||
| BC099529.1 | - | 11 - 12 | 1332 | 137 | ||
| BC132195.1 | - | 11 - 12 | 1297 | 40130472 | 137 | |
| BC138153.1 | - | 11 - 12 | 1297 | 8861172 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013382.1 transcripts.
Set Membership: |
|

Ufl1
Genomic Map