Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 12 - 13 | 1465 | 78 | |
| GenBank mRNA | AK007993.2 | - | 12 - 13 | 1410 | 78 | |
| AK013382.1 | - | 5 - 6 | 727 | 78 | ||
| AK032663.1 | - | 12 - 13 | 1401 | 78 | ||
| AK076206.1 | - | 12 - 13 | 1406 | 78 | ||
| AK082785.1 | - | 2 - 3 | 172 | 78 | ||
| AK129211.1 | - | 11 - 12 | 1378 | 78 | ||
| AK132697.1 | - | 12 - 13 | 1429 | 78 | ||
| BC058809.1 | - | 11 - 12 | 2789 | 78 | ||
| BC099529.1 | - | 12 - 13 | 1452 | 78 | ||
| BC132195.1 | - | 12 - 13 | 1417 | 40130472 | 78 | |
| BC138153.1 | - | 12 - 13 | 1417 | 8861172 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013382.1 transcripts.
Set Membership: |
|

Ufl1
Genomic Map