Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 16 - 17 | 1967 | 141 | |
| GenBank mRNA | AK007993.2 | - | 16 - 17 | 1912 | 141 | |
| AK013382.1 | - | 9 - 10 | 1229 | 141 | ||
| AK032663.1 | - | 16 - 17 | 1903 | 141 | ||
| AK082785.1 | - | 6 - 7 | 674 | 141 | ||
| AK129211.1 | - | 15 - 16 | 1880 | 141 | ||
| AK132697.1 | - | 16 - 17 | 1931 | 141 | ||
| BC058809.1 | - | 15 - 16 | 3291 | 141 | ||
| BC099529.1 | - | 16 - 17 | 1954 | 141 | ||
| BC132195.1 | - | 16 - 17 | 1919 | 40130472 | 141 | |
| BC138153.1 | - | 16 - 17 | 1919 | 8861172 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ufl1
Genomic Map