Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 17 - 18 | 2054 | 74 | |
| GenBank mRNA | AK007993.2 | - | 17 - 18 | 1999 | 74 | |
| AK013382.1 | - | 10 - 11 | 1316 | 74 | ||
| AK032663.1 | - | 17 - 18 | 1990 | 74 | ||
| AK082785.1 | - | 7 - 8 | 761 | 74 | ||
| AK129211.1 | - | 16 - 17 | 1967 | 74 | ||
| AK132697.1 | - | 17 - 18 | 2018 | 74 | ||
| BC058809.1 | - | 16 - 17 | 3378 | 74 | ||
| BC099529.1 | - | 17 - 18 | 2041 | 74 | ||
| BC132195.1 | - | 17 - 18 | 2006 | 40130472 | 74 | |
| BC138153.1 | - | 17 - 18 | 2006 | 8861172 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ufl1
Genomic Map