Assay Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026194.4 | NP_080470.2 | 8 - 9 | 867 | 104 | |
| GenBank mRNA | AK007993.2 | - | 8 - 9 | 812 | 104 | |
| AK013382.1 | - | 1 - 2 | 130 | 104 | ||
| AK032663.1 | - | 8 - 9 | 803 | 104 | ||
| AK076206.1 | - | 8 - 9 | 812 | 104 | ||
| AK129211.1 | - | 7 - 8 | 780 | 104 | ||
| AK132697.1 | - | 8 - 9 | 831 | 104 | ||
| BC058809.1 | - | 7 - 8 | 2191 | 104 | ||
| BC080203.1 | - | 8 - 9 | 845 | 104 | ||
| BC099529.1 | - | 8 - 9 | 854 | 104 | ||
| BC132195.1 | - | 8 - 9 | 819 | 40130472 | 104 | |
| BC138153.1 | - | 8 - 9 | 819 | 8861172 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013382.1 transcripts.
Set Membership: |
|

Ufl1
Genomic Map