Assay Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019992.3 | NP_064376.1 | 7 - 8 | 746 | 94 | |
| GenBank mRNA | AB036058.1 | - | 7 - 8 | 736 | 94 | |
| AK041474.1 | - | 7 - 8 | 747 | 94 | ||
| AK153661.1 | - | 7 - 8 | 825 | 94 | ||
| AK162655.1 | - | 7 - 8 | 828 | 94 | ||
| BC145550.1 | - | 6 - 7 | 648 | 9054114 | 94 | |
| BC145828.1 | - | 7 - 8 | 761 | 40131131 | 94 | |
| BC145830.1 | - | 7 - 8 | 762 | 40131133 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stap1
Genomic Map