Assay Details
| Gene Symbol: Nes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016701.3 | NP_057910.3 | 4 | 2491 | 70 | |
| GenBank mRNA | AF076623.2 | - | 5 | 2323 | 70 | |
| BC060693.1 | - | 4 | 2474 | 6410072 | 70 | |
| BC062893.1 | - | 4 | 2491 | 6405026 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nes
Genomic Map