Assay Details
| Gene Symbol: Pcdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018764.2 | NP_061234.2 | 1 - 2 | 4256 | 80 | |
| GenBank mRNA | AB006758.1 | - | 1 - 2 | 4176 | 80 | |
| AK140356.1 | - | 1 - 2 | 4197 | 80 | ||
| AK159991.1 | - | 1 - 2 | 3187 | 80 | ||
| BC131967.1 | - | 1 - 2 | 3972 | 40130244 | 80 | |
More Information
Set Membership: |
|

Pcdh7
Genomic Map