Assay Details
| Gene Symbol: Tbc1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289514.1 | NP_001276443.1 | 7 - 8 | 1152 | 136 | |
| NM_019636.3 | NP_062610.2 | 8 - 9 | 1733 | 136 | ||
| GenBank mRNA | AK122445.1 | - | 8 - 9 | 2542 | 136 | |
| AK135665.1 | - | 7 - 8 | 137 | |||
| AK135892.1 | - | 7 - 8 | 1417 | 136 | ||
| AK148356.1 | - | 7 - 8 | 1109 | 136 | ||
| AK154591.1 | - | 9 - 10 | 1769 | 136 | ||
| AK159256.1 | - | 8 - 9 | 1729 | 136 | ||
| AK164923.1 | - | 4 - 5 | 437 | 136 | ||
| BC059824.1 | - | 9 - 10 | 2098 | 136 | ||
| BC117931.1 | - | 7 - 8 | 1401 | 40097712 | 136 | |
| U33005.1 | - | 8 - 9 | 136 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d1
Genomic Map