Assay Details
| Gene Symbol: Pcmtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183028.3 | NP_898849.1 | 4 - 5 | 991 | 111 | |
| GenBank mRNA | AK040848.1 | - | 4 - 5 | 719 | 111 | |
| AK079468.1 | - | 3 - 4 | 528 | 111 | ||
| AK145008.1 | - | 4 - 5 | 717 | 111 | ||
| AK161921.1 | - | 1 - 2 | 976 | 111 | ||
| AK166166.1 | - | 4 - 5 | 876 | 111 | ||
| AK166593.1 | - | 4 - 5 | 717 | 111 | ||
| AK168606.1 | - | 4 - 5 | 743 | 111 | ||
| AK168705.1 | - | 4 - 5 | 761 | 111 | ||
| BC019193.1 | - | 4 - 5 | 739 | 111 | ||
| BC052741.1 | - | 3 - 4 | 708 | 6833458 | 111 | |
| BC064801.1 | - | 3 - 4 | 588 | 30067935 | 111 | |
| BC110360.1 | - | 4 - 5 | 761 | 5326388 | 111 | |
| BC117941.1 | - | 3 - 4 | 708 | 40097836 | 111 | |
| BC117942.1 | - | 3 - 4 | 708 | 40097839 | 111 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcmtd1
Genomic Map