Assay Details
| Gene Symbol: Pls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033210.3 | NP_001028382.1 | 1 - 2 | 110 | 129 | |
| GenBank mRNA | AI526519.1 | - | 1 - 2 | 110 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001033210.3 transcripts.
Set Membership: |
|

Pls1
Genomic Map