Assay Details
| Gene Symbol: Slc9a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177909.5 | NP_808577.3 | 10 - 11 | 1372 | 87 | |
| GenBank mRNA | AK037058.1 | - | 10 - 11 | 1356 | 87 | |
| AK156494.1 | - | 10 - 11 | 1333 | 87 | ||
| AK172687.1 | - | 10 - 11 | 1344 | 87 | ||
| BC099942.1 | - | 10 - 11 | 1346 | 30103308 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a9
Genomic Map