Assay Details
| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011206.2 | NP_035336.2 | 4 - 5 | 448 | 72 | |
| GenBank mRNA | AK144353.1 | - | 4 - 5 | 430 | 72 | |
| BC008512.1 | - | 4 - 5 | 418 | 3591418 | 72 | |
| U35124.1 | - | 4 - 5 | 422 | 72 | ||
| U49853.1 | - | 4 - 5 | 388 | 72 | ||
| U52523.1 | - | 4 - 5 | 436 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn18
Genomic Map