Assay Details
| Gene Symbol: Rab23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159729.1 | NP_001153201.1 | 3 - 4 | 467 | 82 | |
| NM_008999.4 | NP_033025.2 | 3 - 4 | 579 | 82 | ||
| GenBank mRNA | AB232618.1 | - | 1 - 2 | 144 | 82 | |
| AK016493.1 | - | 3 - 4 | 479 | 82 | ||
| AK045454.1 | - | 3 - 4 | 338 | 82 | ||
| AK047068.1 | - | 3 - 4 | 580 | 82 | ||
| AK163909.1 | - | 3 - 4 | 588 | 82 | ||
| BC025578.1 | - | 3 - 4 | 474 | 5323566 | 82 | |
| Z22821.1 | - | 1 - 2 | 164 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab23
Genomic Map