Assay Details
| Gene Symbol: Crispld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031402.2 | NP_113579.2 | 3 - 4 | 782 | 91 | |
| GenBank mRNA | AF329198.1 | - | 3 - 4 | 779 | 91 | |
| AK132405.1 | - | 3 - 4 | 520 | 91 | ||
| AK133870.1 | - | 3 - 4 | 783 | 91 | ||
| AK134273.1 | - | 3 - 4 | 718 | 91 | ||
| AK137611.1 | - | 3 - 4 | 783 | 91 | ||
| BC040768.1 | - | 3 - 4 | 1111 | 4458200 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crispld1
Genomic Map