Assay Details
| Gene Symbol: Stard8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199018.2 | NP_950183.1 | 6 - 7 | 2017 | 109 | |
| GenBank mRNA | AK133927.1 | - | 6 - 7 | 1965 | 109 | |
| AK149830.1 | - | 6 - 7 | 1948 | 109 | ||
| AK150259.1 | - | 6 - 7 | 1948 | 109 | ||
| BC034186.1 | - | 2 - 3 | 1139 | 109 | ||
| BC062944.1 | - | 6 - 7 | 1932 | 30357177 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stard8
Genomic Map