Assay Details
| Gene Symbol: Rab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034408.1 | - | 1 - 2 | 209 | 140 | |
| AK050526.1 | - | 1 - 2 | 205 | 140 | ||
| AK129477.1 | - | 1 - 2 | 199 | 140 | ||
| AK151085.1 | - | 1 - 2 | 205 | 140 | ||
| AK152978.1 | - | 1 - 2 | 210 | 140 | ||
| AK159425.1 | - | 1 - 2 | 205 | 140 | ||
| AK166249.1 | - | 1 - 2 | 202 | 131 | ||
| AK167150.1 | - | 1 - 2 | 209 | 140 | ||
| BC002077.1 | - | 1 - 2 | 208 | 3592802 | 140 | |
| Y00094.1 | - | 1 - 2 | 142 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034408.1, AK151085.1, AK159425.1, AK166249.1, AK167150.1, BC002077.1, Y00094.1 transcripts.
Set Membership: |
|

Rab1
Genomic Map