Assay Details
| Gene Symbol: Rab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008996.3 | NP_033022.1 | 3 - 4 | 227 | 140 | |
| GenBank mRNA | AB232583.1 | - | 3 - 4 | 199 | 140 | |
| AF226873.1 | - | 3 - 4 | 227 | 140 | ||
| AK034408.1 | - | 3 - 4 | 395 | 140 | ||
| AK151085.1 | - | 3 - 4 | 140 | |||
| AK152978.1 | - | 3 - 4 | 396 | 140 | ||
| AK159425.1 | - | 3 - 4 | 391 | 140 | ||
| AK166249.1 | - | 3 - 4 | 388 | 140 | ||
| AK167150.1 | - | 3 - 4 | 395 | 140 | ||
| AK197899.1 | - | 286 | 140 | |||
| BC002077.1 | - | 3 - 4 | 394 | 3592802 | 140 | |
| Y00094.1 | - | 3 - 4 | 328 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab1
Genomic Map