Assay Details
| Gene Symbol: Rab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008996.3 | NP_033022.1 | 4 - 5 | 316 | 72 | |
| GenBank mRNA | AB232583.1 | - | 4 - 5 | 288 | 72 | |
| AF226873.1 | - | 4 - 5 | 316 | 72 | ||
| AK034408.1 | - | 4 - 5 | 484 | 72 | ||
| AK129477.1 | - | 4 - 5 | 474 | 72 | ||
| AK151085.1 | - | 4 - 5 | 480 | 72 | ||
| AK152978.1 | - | 4 - 5 | 485 | 72 | ||
| AK159425.1 | - | 4 - 5 | 480 | 72 | ||
| AK166249.1 | - | 4 - 5 | 477 | 72 | ||
| AK167150.1 | - | 4 - 5 | 484 | 72 | ||
| BC002077.1 | - | 4 - 5 | 483 | 3592802 | 72 | |
| Y00094.1 | - | 4 - 5 | 417 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab1
Genomic Map