Assay Details
| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015794.1 | NP_056609.1 | 4 - 5 | 2068 | 79 | |
| GenBank mRNA | AF176529.1 | - | 4 - 5 | 788 | 79 | |
| AK154497.1 | - | 4 - 5 | 577 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl17
Genomic Map