Assay Details
| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015794.1 | NP_056609.1 | 5 - 6 | 2182 | 80 | |
| GenBank mRNA | AF176529.1 | - | 5 - 6 | 902 | 80 | |
| AK154497.1 | - | 5 - 6 | 691 | 80 | ||
| BC058120.1 | - | 2 - 3 | 240 | 6810624 | 80 | |
| BC138330.1 | - | 2 - 3 | 218 | 8861350 | 80 | |
| BC138331.1 | - | 2 - 3 | 218 | 8861351 | 80 | |
More Information
Set Membership: |
|

Fbxl17
Genomic Map