Assay Details
| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015794.1 | NP_056609.1 | 7 - 8 | 2386 | 70 | |
| GenBank mRNA | AF176529.1 | - | 7 - 8 | 1106 | 70 | |
| AK004419.1 | - | 1 - 2 | 68 | 70 | ||
| AK154497.1 | - | 7 - 8 | 895 | 70 | ||
| BC058120.1 | - | 4 - 5 | 444 | 6810624 | 70 | |
| BC138330.1 | - | 4 - 5 | 422 | 8861350 | 70 | |
| BC138331.1 | - | 4 - 5 | 422 | 8861351 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl17
Genomic Map