Assay Details
| Gene Symbol: Slc1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018861.3 | NP_061349.3 | 6 - 7 | 1471 | 88 | |
| GenBank mRNA | AF246129.1 | - | 6 - 7 | 1230 | 88 | |
| AK044280.1 | - | 6 - 7 | 1442 | 88 | ||
| AK136900.1 | - | 6 - 7 | 1471 | 88 | ||
| AK139248.1 | - | 6 - 7 | 1470 | 88 | ||
| AK140866.1 | - | 6 - 7 | 1472 | 88 | ||
| AK148075.1 | - | 6 - 7 | 1472 | 88 | ||
| BC043483.1 | - | 6 - 7 | 1429 | 5003744 | 88 | |
| BC052733.1 | - | 6 - 7 | 1345 | 6816689 | 88 | |
| U75215.1 | - | 6 - 7 | 1341 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a4
Genomic Map