Assay Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY753561.1 | - | 2 - 3 | 172 | 67 | |
| AY753562.1 | - | 2 - 3 | 172 | 67 | ||
| DQ025533.1 | - | 2 - 3 | 172 | 67 | ||
More Information
Set Membership: |
|

Vezt
Genomic Map