Assay Details
| Gene Symbol: Metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019648.3 | NP_062622.1 | 5 - 6 | 1118 | 77 | |
| GenBank mRNA | AB003144.1 | - | 4 - 5 | 429 | 77 | |
| AF434712.1 | - | 4 - 5 | 508 | 77 | ||
| AK076804.1 | - | 4 - 5 | 461 | 77 | ||
| AK090323.1 | - | 4 - 5 | 449 | 77 | ||
| AK147097.1 | - | 4 - 5 | 553 | 77 | ||
| AK159650.1 | - | 4 - 5 | 533 | 77 | ||
| AK163318.1 | - | 4 - 5 | 535 | 77 | ||
| AK165301.1 | - | 5 - 6 | 1120 | 77 | ||
| BC002213.1 | - | 4 - 5 | 530 | 3490341 | 77 | |
| BC010263.1 | - | 4 - 5 | 503 | 77 | ||
| BC092052.1 | - | 4 - 5 | 495 | 3670685 | 77 | |
More Information
Set Membership: |
|

Metap2
Genomic Map