Assay Details
| Gene Symbol: Metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019648.3 | NP_062622.1 | 6 - 7 | 1284 | 88 | |
| GenBank mRNA | AB003144.1 | - | 5 - 6 | 595 | 88 | |
| AF434712.1 | - | 5 - 6 | 674 | 88 | ||
| AK076804.1 | - | 5 - 6 | 627 | 88 | ||
| AK090323.1 | - | 5 - 6 | 615 | 88 | ||
| AK147097.1 | - | 5 - 6 | 719 | 88 | ||
| AK159650.1 | - | 5 - 6 | 698 | 88 | ||
| AK163318.1 | - | 5 - 6 | 701 | 88 | ||
| AK165301.1 | - | 6 - 7 | 1286 | 88 | ||
| AK165845.1 | - | 5 - 6 | 610 | 88 | ||
| BC002213.1 | - | 5 - 6 | 696 | 3490341 | 88 | |
| BC010263.1 | - | 5 - 6 | 668 | 88 | ||
| BC092052.1 | - | 5 - 6 | 661 | 3670685 | 88 | |
More Information
Set Membership: |
|

Metap2
Genomic Map