Assay Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029852.2 | NP_084128.2 | 10 - 11 | 1696 | 163 | |
| GenBank mRNA | AK015007.1 | - | 10 - 11 | 1833 | 163 | |
| AK135868.1 | - | 2 - 3 | 193 | 163 | ||
| AK151291.1 | - | 4 - 5 | 545 | 163 | ||
| AK152465.1 | - | 4 - 5 | 545 | 163 | ||
| AK152801.1 | - | 4 - 5 | 545 | 163 | ||
| AK153515.1 | - | 4 - 5 | 545 | 163 | ||
| BC028960.1 | - | 9 - 10 | 1397 | 163 | ||
| BC043468.1 | - | 9 - 10 | 1310 | 163 | ||
| BC070464.1 | - | 10 - 11 | 1696 | 30608497 | 163 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cep83
Genomic Map