Assay Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029852.2 | NP_084128.2 | 11 - 12 | 1772 | 122 | |
| GenBank mRNA | AK015007.1 | - | 11 - 12 | 1909 | 122 | |
| AK135868.1 | - | 3 - 4 | 269 | 122 | ||
| AK151291.1 | - | 5 - 6 | 621 | 122 | ||
| AK152465.1 | - | 5 - 6 | 621 | 122 | ||
| AK152801.1 | - | 5 - 6 | 621 | 122 | ||
| AK153515.1 | - | 5 - 6 | 621 | 122 | ||
| BC028960.1 | - | 10 - 11 | 1473 | 122 | ||
| BC043468.1 | - | 10 - 11 | 1386 | 122 | ||
| BC070464.1 | - | 11 - 12 | 1772 | 30608497 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cep83
Genomic Map