Assay Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029852.2 | NP_084128.2 | 12 - 13 | 1931 | 81 | |
| GenBank mRNA | AK015007.1 | - | 12 - 13 | 2068 | 81 | |
| AK135868.1 | - | 4 - 5 | 428 | 81 | ||
| AK151291.1 | - | 6 - 7 | 780 | 81 | ||
| AK152465.1 | - | 6 - 7 | 780 | 81 | ||
| AK152801.1 | - | 6 - 7 | 780 | 81 | ||
| AK153515.1 | - | 6 - 7 | 780 | 81 | ||
| BC028960.1 | - | 11 - 12 | 1632 | 81 | ||
| BC043468.1 | - | 11 - 12 | 1545 | 81 | ||
| BC070464.1 | - | 12 - 13 | 1931 | 30608497 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cep83
Genomic Map