Assay Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029852.2 | NP_084128.2 | 1 - 2 | 257 | 138 | |
| GenBank mRNA | AK011135.1 | - | 1 - 2 | 329 | 138 | |
| AK015007.1 | - | 1 - 2 | 394 | 138 | ||
| AK141627.1 | - | 1 - 2 | 378 | 138 | ||
| BC070464.1 | - | 1 - 2 | 257 | 30608497 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015007.1, AK141627.1, BC070464.1, NM_029852.2 transcripts.
Set Membership: |
|

Cep83
Genomic Map