Assay Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029852.2 | NP_084128.2 | 2 - 3 | 527 | 131 | |
| GenBank mRNA | AK011135.1 | - | 2 - 3 | 599 | 131 | |
| AK015007.1 | - | 2 - 3 | 664 | 131 | ||
| AK141627.1 | - | 2 - 3 | 648 | 131 | ||
| BC028960.1 | - | 1 - 2 | 228 | 131 | ||
| BC043468.1 | - | 1 - 2 | 141 | 131 | ||
| BC070464.1 | - | 2 - 3 | 527 | 30608497 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cep83
Genomic Map