Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00002424.mm.1 |
NM_001040089.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002424.mm.1 |
NM_001040089.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002424.mm.1 |
TC0X00000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002425.mm.1 |
NM_001040089.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002425.mm.1 |
NM_001040089.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002425.mm.1 |
TC0X00000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002426.mm.1 |
NM_001040089.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002426.mm.1 |
NM_001040089.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002426.mm.1 |
TC0X00000353.mm.1 |
MTA TC |