Assay Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008777.3 | NP_032803.2 | 8 - 9 | 1130 | 95 | |
| GenBank mRNA | BC013458.1 | - | 8 - 9 | 988 | 4223155 | 95 |
| X51942.1 | - | 8 - 9 | 955 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pah
Genomic Map