Assay Details
| Gene Symbol: Gm1123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080776.1 | NP_001074245.1 | 4 - 5 | 654 | 79 | |
| GenBank mRNA | AB262662.1 | - | 4 - 5 | 683 | 79 | |
| BC114986.1 | - | 4 - 5 | 654 | 40056015 | 79 | |
| BC115956.1 | - | 4 - 5 | 654 | 40056016 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gm1123
Genomic Map