Assay Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029094.3 | NP_083370.2 | 12 - 13 | 1912 | 76 | |
| GenBank mRNA | AK090116.1 | - | 12 - 13 | 1789 | 76 | |
| AK154106.1 | - | 12 - 13 | 1897 | 76 | ||
| BC127073.1 | - | 11 - 12 | 1561 | 40110218 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3cb
Genomic Map