Assay Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029094.3 | NP_083370.2 | 19 - 20 | 2837 | 73 | |
| GenBank mRNA | AK003230.1 | - | 3 - 4 | 252 | 73 | |
| AK081350.1 | - | 19 - 20 | 3028 | 73 | ||
| AK090116.1 | - | 19 - 20 | 2714 | 73 | ||
| AK154106.1 | - | 19 - 20 | 2822 | 73 | ||
| AK165230.1 | - | 297 | 73 | |||
| BC127073.1 | - | 18 - 19 | 2486 | 40110218 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3cb
Genomic Map