Assay Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029094.3 | NP_083370.2 | 7 - 8 | 1304 | 134 | |
| GenBank mRNA | AK081350.1 | - | 7 - 8 | 1499 | 134 | |
| AK090116.1 | - | 7 - 8 | 1181 | 134 | ||
| AK154106.1 | - | 7 - 8 | 1289 | 134 | ||
| BC039650.1 | - | 7 - 8 | 1267 | 134 | ||
| BC127073.1 | - | 6 - 7 | 953 | 40110218 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3cb
Genomic Map