Assay Details
| Gene Symbol: Dis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028315.2 | NP_082591.2 | 10 - 11 | 1600 | 65 | |
| GenBank mRNA | AK032091.1 | - | 5 - 6 | 690 | 65 | |
| AK038313.1 | - | 10 - 11 | 1470 | 65 | ||
| AK129259.1 | - | 6 - 7 | 2597 | 65 | ||
| BC027357.1 | - | 10 - 11 | 1570 | 4952415 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dis3
Genomic Map