Assay Details
| Gene Symbol: Dis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028315.2 | NP_082591.2 | 6 - 7 | 1084 | 76 | |
| GenBank mRNA | AK032091.1 | - | 1 - 2 | 174 | 76 | |
| AK038313.1 | - | 6 - 7 | 954 | 76 | ||
| AK129259.1 | - | 3 - 4 | 1372 | 76 | ||
| BC027357.1 | - | 6 - 7 | 1054 | 4952415 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dis3
Genomic Map