Assay Details
| Gene Symbol: Bora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175265.4 | NP_780474.1 | 4 - 5 | 618 | 101 | |
| GenBank mRNA | AK034930.1 | - | 4 - 5 | 619 | 101 | |
| AK141389.1 | - | 4 - 5 | 569 | 101 | ||
| AK167430.1 | - | 4 - 5 | 550 | 101 | ||
| BC052183.1 | - | 4 - 5 | 519 | 6336038 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bora
Genomic Map