Assay Details
| Gene Symbol: Maged1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019791.2 | NP_062765.1 | 1 - 2 | 180 | 66 | |
| GenBank mRNA | AB029448.1 | - | 1 - 2 | 180 | 66 | |
| AK017275.1 | - | 1 - 2 | 98 | 66 | ||
| AK049660.1 | - | 1 - 2 | 131 | 66 | ||
| AK087937.1 | - | 1 - 2 | 131 | 66 | ||
| AK132248.1 | - | 1 - 2 | 119 | 66 | ||
| AK135452.1 | - | 1 - 2 | 130 | 66 | ||
| AK145872.1 | - | 1 - 2 | 100 | 66 | ||
| AK146867.1 | - | 1 - 2 | 130 | 66 | ||
| AK146908.1 | - | 1 - 2 | 130 | 66 | ||
| AK220150.1 | - | 1 - 2 | 109 | 66 | ||
| BC016438.1 | - | 1 - 2 | 96 | 4480947 | 66 | |
| BC031461.1 | - | 1 - 2 | 120 | 4018875 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB029448.1, AK017275.1, AK049660.1, AK132248.1, AK135452.1, AK145872.1, AK146867.1, AK146908.1, BC016438.1, BC031461.1, NM_019791.2 transcripts.
Set Membership: |
|

Maged1
Genomic Map